Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028726.hg.1 |
TC05002093.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028726.hg.1 |
ENST00000310418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028726.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000507821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000507821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000509319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000509319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000509319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190048.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190048.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190048.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190048.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190048.hg.1 |
TC0500012938.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000310418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000345807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000502877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000507821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000507821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000508425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000509319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000509319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000509319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
ENST00000510734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_001834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NM_007097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012938.hg.1 |
NR_045724.1 |
Refseq |