Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK304443.1 | - | 22 - 23 | 2784 | 84 | |
AK316511.1 | - | 22 - 23 | 3218 | 84 |
Gene Symbol: LOC107984125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017014157.1 | XP_016869646.1 | 15 - 16 | 2331 | 84 | |
XM_017014158.1 | XP_016869647.1 | 10 - 11 | 1667 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |