Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010859.1 | NP_001010859.1 | 4 - 5 | 440 | 145 | |
XM_011529922.2 | XP_011528224.1 | 5 - 6 | 939 | 145 | ||
XM_017028629.1 | XP_016884118.1 | 5 - 6 | 473 | 145 | ||
XM_017028631.1 | XP_016884120.1 | 5 - 6 | 952 | 145 | ||
GenBank mRNA | CR456485.1 | - | 4 - 5 | 440 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |