Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291815.1 | NP_001278744.1 | 66 - 67 | 10140 | 107 | |
XM_011518465.2 | XP_011516767.1 | 66 - 67 | 10277 | 107 | ||
XM_011518466.2 | XP_011516768.1 | 66 - 67 | 10277 | 107 | ||
XM_011518467.2 | XP_011516769.1 | 65 - 66 | 10100 | 107 | ||
XM_011518468.2 | XP_011516770.1 | 66 - 67 | 10277 | 107 | ||
XM_011518469.2 | XP_011516771.1 | 66 - 67 | 10277 | 107 | ||
XM_017014585.1 | XP_016870074.1 | 45 - 46 | 6946 | 107 | ||
XM_017014586.1 | XP_016870075.1 | 20 - 21 | 2874 | 107 | ||
GenBank mRNA | AL834139.1 | - | 7 - 8 | 853 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |