Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016060.hg.1 |
TC0X001189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016060.hg.1 |
ENST00000332921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016060.hg.1 |
ENST00000395409.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016060.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016060.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016059.hg.1 |
TC0X001189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016059.hg.1 |
ENST00000332921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016059.hg.1 |
ENST00000395409.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016059.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016059.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016062.hg.1 |
TC0X001189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016062.hg.1 |
ENST00000332921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016062.hg.1 |
ENST00000395409.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016062.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016062.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169622.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169622.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169622.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169622.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169622.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169622.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169622.hg.1 |
TC0X00010204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010204.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169623.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169623.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169623.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169623.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169623.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169623.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169623.hg.1 |
TC0X00010204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00169625.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169625.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169625.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169625.hg.1 |
ENST00000395409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169625.hg.1 |
ENST00000509231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169625.hg.1 |
NM_001012980.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169625.hg.1 |
TC0X00010204.hg.1 |
Clariom D TC |