Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011527471.2 | XP_011525773.1 | 30 - 31 | 3743 | 150 | |
XM_011527472.2 | XP_011525774.1 | 30 - 31 | 3728 | 150 | ||
XM_011527473.2 | XP_011525775.1 | 30 - 31 | 3743 | 150 | ||
XM_011527474.2 | XP_011525776.1 | 29 - 30 | 3623 | 150 | ||
XM_011527475.2 | XP_011525777.1 | 30 - 31 | 3743 | 150 | ||
XM_017027458.1 | XP_016882947.1 | 30 - 31 | 3743 | 150 | ||
XM_017027459.1 | XP_016882948.1 | 20 - 21 | 2726 | 150 | ||
GenBank mRNA | AK023134.1 | - | 4 - 5 | 538 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |