Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010518.hg.1 |
TC02000670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010518.hg.1 |
ENST00000309415.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010518.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010519.hg.1 |
TC02000670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010519.hg.1 |
ENST00000309415.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010519.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010520.hg.1 |
TC02000670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010520.hg.1 |
ENST00000309415.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010520.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010517.hg.1 |
TC02000670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010517.hg.1 |
ENST00000309415.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010517.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161601.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161601.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161601.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161601.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161601.hg.1 |
TC0200008811.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008811.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161602.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161602.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161602.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161602.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161602.hg.1 |
TC0200008811.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200161597.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161597.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161597.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161597.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161597.hg.1 |
TC0200008811.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200161600.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161600.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161600.hg.1 |
ENST00000309415.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161600.hg.1 |
NM_001099289.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161600.hg.1 |
TC0200008811.hg.1 |
Clariom D TC |