Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000533217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000533217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000533217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
ENST00000533217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000351018.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000396979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000533217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000533217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000533217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
ENST00000533217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009864.hg.1 |
NM_001665.3 |
Refseq |