Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005876.4 | NP_005867.3 | 24 - 25 | 5169 | 95 | |
XM_005246237.2 | XP_005246294.1 | 26 - 27 | 4897 | 95 | ||
XM_005246239.2 | XP_005246296.1 | 26 - 27 | 4712 | 95 | ||
XM_005246240.2 | XP_005246297.1 | 19 - 20 | 2914 | 95 | ||
XM_005246241.1 | XP_005246298.1 | 19 - 20 | 2808 | 95 | ||
XM_005246242.4 | XP_005246299.1 | 19 - 20 | 2933 | 95 | ||
XM_006712189.3 | XP_006712252.1 | 25 - 26 | 4902 | 95 | ||
XM_006712193.3 | XP_006712256.1 | 19 - 20 | 6033 | 95 | ||
XM_011510479.2 | XP_011508781.1 | 25 - 26 | 5335 | 95 | ||
XM_011510483.2 | XP_011508785.2 | 25 - 26 | 4932 | 95 | ||
XM_017003157.1 | XP_016858646.1 | 25 - 26 | 4932 | 95 | ||
XM_017003158.1 | XP_016858647.1 | 21 - 22 | 4508 | 95 | ||
XM_017003159.1 | XP_016858648.1 | 22 - 23 | 4691 | 95 | ||
XM_017003160.1 | XP_016858649.1 | 9 - 10 | 1104 | 95 | ||
GenBank mRNA | AB037718.1 | - | 15 - 16 | 2226 | 95 | |
AY603755.1 | - | 23 - 24 | 4782 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Homo sapiens Probe spans exons |