Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012290.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012290.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012290.hg.1 |
TC11000984.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012291.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012291.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012291.hg.1 |
TC11000984.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012292.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012292.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012292.hg.1 |
TC11000984.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012288.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012288.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012288.hg.1 |
TC11000984.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012289.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012289.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012289.hg.1 |
TC11000984.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012293.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012293.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012293.hg.1 |
TC11000984.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100166514.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166514.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166514.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166514.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166514.hg.1 |
TC1100008994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100166518.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166518.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166518.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166518.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166518.hg.1 |
TC1100008994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100166519.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166519.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166519.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166519.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166519.hg.1 |
TC1100008994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100166516.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166516.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166516.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166516.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166516.hg.1 |
TC1100008994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100166515.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166515.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166515.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166515.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166515.hg.1 |
TC1100008994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100166517.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166517.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166517.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166517.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166517.hg.1 |
TC1100008994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100166520.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166520.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166520.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166520.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166520.hg.1 |
TC1100008994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
ENST00000327419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008994.hg.1 |
NM_207645.3 |
Refseq |