Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8118727 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118730 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118731 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118729 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118733 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118728 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8118732 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8118726 |
- |
- |
Affymetrix Human Genome U219 Array |
11750339_at |
- |
- |
Affymetrix Human Genome U219 Array |
11750339_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005088.hg.1 |
TC06000426.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005088.hg.1 |
ENST00000374458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005088.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005089.hg.1 |
TC06000426.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005089.hg.1 |
ENST00000374458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005089.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000610663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000616119.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014116.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198528.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198528.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198528.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198528.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198528.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198528.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198528.hg.1 |
TC0600014116.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
ENST00000374458.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
ENST00000613113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
ENST00000618642.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
NR_028361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198529.hg.1 |
TC0600014116.hg.1 |
Clariom D TC |