Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014693.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014693.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014693.hg.1 |
TC11001162.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014694.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014694.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014694.hg.1 |
TC11001162.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009427.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170853.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170853.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170853.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170853.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170853.hg.1 |
TC1100009427.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100170854.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170854.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170854.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170854.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170854.hg.1 |
TC1100009427.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100170852.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170852.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170852.hg.1 |
ENST00000445202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170852.hg.1 |
NM_001129883.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170852.hg.1 |
TC1100009427.hg.1 |
Clariom D TC |