Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105079.2 | NP_001098549.2 | 3 - 4 | 674 | 64 | |
XM_011545916.2 | XP_011544218.1 | 3 - 4 | 2051 | 64 | ||
GenBank mRNA | DB082271.1 | - | 3 - 4 | 271 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons |