Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009464.hg.1 |
TC0X000735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009464.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009464.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159380.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159380.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159380.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159380.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159380.hg.1 |
TC0X00008737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
ENST00000329903.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008737.hg.1 |
NM_005140.1 |
Refseq |