Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198318.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198318.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198318.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198318.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198318.hg.1 |
TC0100013264.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013264.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198315.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198315.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198315.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198315.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198315.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198315.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198315.hg.1 |
TC0100013264.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198316.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198316.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198316.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198316.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198316.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198316.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198316.hg.1 |
TC0100013264.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198317.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198317.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198317.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198317.hg.1 |
ENST00000426928.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198317.hg.1 |
ENST00000618559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198317.hg.1 |
NM_001289974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198317.hg.1 |
TC0100013264.hg.1 |
Clariom D TC |