Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015255.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015255.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015255.hg.1 |
TC14001343.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400172163.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172163.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172163.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172163.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172163.hg.1 |
TC1400009778.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
ENST00000216468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009778.hg.1 |
NM_213601.2 |
Refseq |