Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277269.1 | NP_001264198.1 | 29 - 30 | 3717 | 106 | |
NM_001292063.1 | NP_001278992.1 | 30 - 31 | 3681 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |