Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL137642.1 | - | 2 - 3 | 388 | 67 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | STEAP2 metalloreductase | STEAP2 |
IPCA1, PCANAP1, PUMPCn, STAMP1, STMP |
-82.7kb | Sense | Chr. 7 - 90211686 - 90243408 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101927446 | LOC101927446 | 12.6kb | Sense | Chr. 7 - 90335223 - 90344094 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |