Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207437.3 | NP_997320.2 | 45 - 46 | 7610 | 80 | |
XM_005253555.3 | XP_005253612.1 | 46 - 47 | 8273 | 80 | ||
XM_011538014.2 | XP_011536316.1 | 45 - 46 | 8114 | 80 | ||
XM_011538015.2 | XP_011536317.1 | 45 - 46 | 8084 | 80 | ||
XM_011538016.2 | XP_011536318.1 | 46 - 47 | 8273 | 80 | ||
XM_011538017.2 | XP_011536319.1 | 44 - 45 | 7937 | 80 | ||
XM_011538019.2 | XP_011536321.1 | 28 - 29 | 4872 | 80 | ||
XM_011538020.2 | XP_011536322.1 | 45 - 46 | 8135 | 80 | ||
XM_017018960.1 | XP_016874449.1 | 46 - 47 | 8273 | 80 | ||
XM_017018961.1 | XP_016874450.1 | 45 - 46 | 7964 | 80 | ||
XM_017018962.1 | XP_016874451.1 | 28 - 29 | 4794 | 80 | ||
GenBank mRNA | AB095937.2 | - | 20 - 21 | 3325 | 80 | |
AB290168.1 | - | 25 - 26 | 4129 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons |