Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8176437 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177105 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8176439 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177107 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177103 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177102 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8176441 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177104 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8176436 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177106 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8176440 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8176438 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8176435 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8177101 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000306.hg.1 |
TC0Y000028.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000306.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000306.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000306.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000306.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000306.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00144599.hg.1 |
TC0Y00006518.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006984.hg.1 |
ENST00000624593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000276770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000449828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
ENST00000457100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_001534.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00006518.hg.1 |
NR_003592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000276779.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000415405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007013.hg.1 |
ENST00000447655.1 |
Ensembl |