Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000550823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000550823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000550823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020142.hg.1 |
ENST00000341809.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020142.hg.1 |
ENST00000537195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020142.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020142.hg.1 |
TC12001533.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200182161.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182161.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182161.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182161.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182161.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182161.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182161.hg.1 |
TC1200010769.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000341809.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000550823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000550823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000550823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
ENST00000553168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010769.hg.1 |
NM_175078.2 |
Refseq |