Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8170646 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8170647 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8170645 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8170644 |
- |
- |
Affymetrix Human Genome U219 Array |
11742142_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11742142_PM_x_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000517530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000522479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009728.hg.1 |
TC0X000748.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009728.hg.1 |
ENST00000423874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009728.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009729.hg.1 |
TC0X000748.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009729.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
ENST00000569962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008780.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000340042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000344692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000517530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000519876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000520021.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000521682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000522479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
ENST00000523812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012374.hg.1 |
NM_138465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159750.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159750.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159750.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159750.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159750.hg.1 |
TC0X00008780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
ENST00000338647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
ENST00000569962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
NM_001136273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008780.hg.1 |
NM_001136273.1 |
Refseq |
Human Genome U133 Set |
233116_at |
- |
- |
Human Genome U133 Set |
233116_PM_at |
- |
- |