Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014510.hg.1 |
TC09001087.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014510.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014510.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900169113.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900169113.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900169113.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900169113.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900169113.hg.1 |
TC0900010052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
ENST00000377694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010052.hg.1 |
NM_001007563.2 |
Refseq |