Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006925.hg.1 |
ENST00000404743.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006925.hg.1 |
ENST00000539337.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006925.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006925.hg.1 |
TC10000630.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008383.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156788.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156788.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156788.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156788.hg.1 |
ENST00000404743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156788.hg.1 |
ENST00000539337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156788.hg.1 |
NM_001128215.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156788.hg.1 |
TC1000008383.hg.1 |
Clariom D TC |