Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
96054 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
51233 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
830940 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
641939 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
AB033033.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
AF182316.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
AF182317.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
AK075258.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
AK093890.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
AL096713.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
BC014612.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
BC052617.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
NM_013451.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
203183 |
NM_133337.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
440615 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
342380 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
379940 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
362287 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
343479 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
71117 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P227930 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_32_P227930 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_2005 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_2006 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
17874 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
32698 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P24643551 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P01374360 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P17363642 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P17363649 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P227930 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_32_P227930 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577649 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398037 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577630 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398057 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577628 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577626 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577635 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398052 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398051 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398058 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577627 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577656 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577659 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398063 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577636 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577623 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398042 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398041 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577645 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15755926 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398044 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398033 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577622 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398054 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398048 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398043 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577656 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398044 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398063 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577628 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577630 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398058 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398052 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398057 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577635 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577649 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398051 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398042 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577636 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398043 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577623 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577627 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398041 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398033 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577622 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15755926 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577645 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398048 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577659 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398054 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P24577626 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P04398037 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_32_P227930 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_32_P227930 |
- |
- |
Agilent Human 1B Microarray G4111A |
A_32_P227930 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398070 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398099 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398083 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398051 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398036 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398101 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398060 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398068 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398090 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15755923 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398077 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15755921 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398096 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398108 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398086 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398112 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398041 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398058 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398054 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398105 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398044 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398097 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398063 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398079 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398074 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398042 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398093 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398048 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398064 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398084 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398110 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398081 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398091 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398033 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398057 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P04398106 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P17363642 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P17363649 |
- |
- |