Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011760.hg.1 |
TC05000822.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011760.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011760.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000509831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000509831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000509831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
ENST00000509831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011758.hg.1 |
TC05000822.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011758.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011758.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011759.hg.1 |
TC05000822.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011759.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011759.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194533.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194533.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194533.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194533.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194533.hg.1 |
TC0500013258.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194536.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194536.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194536.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194536.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194536.hg.1 |
TC0500013258.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000509831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000509831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000509831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
ENST00000509831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013258.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194535.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194535.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194535.hg.1 |
ENST00000333677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194535.hg.1 |
NM_001001669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194535.hg.1 |
TC0500013258.hg.1 |
Clariom D TC |