Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033517.1 | NP_277052.1 | 18 - 19 | 2136 | 112 | |
GenBank mRNA | AB569469.1 | - | 18 - 19 | 2136 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |