Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021626.hg.1 |
TC03001268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021626.hg.1 |
ENST00000282538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021626.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021626.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000498387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000498387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000498387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
ENST00000498387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177326.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177326.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177326.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177326.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177326.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177326.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177326.hg.1 |
TC0300010620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000282538.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000454381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000498387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000498387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000498387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
ENST00000498387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010620.hg.1 |
NM_207359.2 |
Refseq |