Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014393.hg.1 |
TC0X001065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014393.hg.1 |
ENST00000168216.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014393.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014393.hg.1 |
ENST00000375304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014393.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014393.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014393.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014391.hg.1 |
TC0X001065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014391.hg.1 |
ENST00000168216.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014391.hg.1 |
ENST00000375304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014391.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014391.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014391.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166712.hg.1 |
TC0X00009762.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375298.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000477706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009762.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
ENST00000168216.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
ENST00000375304.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
ENST00000495986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
NM_001037811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
NM_004493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00166710.hg.1 |
TC0X00009762.hg.1 |
Clariom D TC |