Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026574.hg.1 |
TC06002119.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026574.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026574.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191139.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191139.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191139.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191139.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191139.hg.1 |
TC0600013203.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
ENST00000367928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013203.hg.1 |
NM_004666.2 |
Refseq |