Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005681.hg.1 |
TC01000368.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005681.hg.1 |
ENST00000373943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005681.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005681.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000481874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000481874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
ENST00000481874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000472285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000481874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000481874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
ENST00000481874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007532.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
ENST00000373943.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
ENST00000440806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
ENST00000468761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
NM_177424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152685.hg.1 |
TC0100007532.hg.1 |
Clariom D TC |