Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013660.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013660.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013660.hg.1 |
TC14001190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000555420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000555420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000555420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000555420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
ENST00000556317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000553903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000555420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000555420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000555420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000555420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
ENST00000556317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009355.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169159.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169159.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169159.hg.1 |
ENST00000261249.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169159.hg.1 |
NM_020810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169159.hg.1 |
TC1400009355.hg.1 |
Clariom D TC |