Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019741.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019741.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019741.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019741.hg.1 |
TC10001557.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
ENST00000506028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011521.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178299.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178299.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178299.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178299.hg.1 |
ENST00000357947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178299.hg.1 |
ENST00000469598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178299.hg.1 |
NM_012465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178299.hg.1 |
TC1000011521.hg.1 |
Clariom D TC |