Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005277.hg.1 |
ENST00000261861.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005277.hg.1 |
ENST00000540068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005277.hg.1 |
ENST00000543950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005277.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005277.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005277.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005277.hg.1 |
TC15000629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154491.hg.1 |
TC1500007677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000261861.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000540068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000543950.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
ENST00000566799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190456.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_001190457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007677.hg.1 |
NM_006091.4 |
Refseq |