Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017538.hg.1 |
ENST00000326053.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017538.hg.1 |
ENST00000525780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017538.hg.1 |
ENST00000528998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017538.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017538.hg.1 |
TC11001391.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017537.hg.1 |
ENST00000326053.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017537.hg.1 |
ENST00000525780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017537.hg.1 |
ENST00000528998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017537.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017537.hg.1 |
TC11001391.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017539.hg.1 |
ENST00000326053.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017539.hg.1 |
ENST00000525780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017539.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017539.hg.1 |
TC11001391.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176105.hg.1 |
TC1100010047.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176101.hg.1 |
TC1100010047.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176103.hg.1 |
TC1100010047.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000326053.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000525780.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000526227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000527607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
ENST00000528998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010047.hg.1 |
NM_020643.2 |
Refseq |