Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023395.hg.1 |
TC08001758.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023395.hg.1 |
ENST00000377307.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023395.hg.1 |
ENST00000528810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023395.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023394.hg.1 |
TC08001758.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023394.hg.1 |
ENST00000377307.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08023394.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000528810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000528810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
ENST00000528810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000528810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000528810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
ENST00000528810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012225.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
ENST00000276826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
ENST00000377307.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
NM_001308207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
NM_001308208.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
NM_025251.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183990.hg.1 |
TC0800012225.hg.1 |
Clariom D TC |