Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009520.hg.1 |
ENST00000396720.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009520.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009520.hg.1 |
TC19000680.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000602258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000602258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000602258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000602258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164784.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164784.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164784.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900164784.hg.1 |
TC1900008428.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900164784.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164784.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900164784.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000396720.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000594866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000602258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000602258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000602258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000602258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
ENST00000614245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008428.hg.1 |
NM_015711.3 |
Refseq |