Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005355.3 | NP_005346.3 | 7 - 8 | 1091 | 134 | |
XM_011535803.2 | XP_011534105.1 | 9 - 10 | 1198 | 134 | ||
GenBank mRNA | BC130489.1 | - | 7 - 8 | 1139 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |