Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017008378.1 | XP_016863867.1 | 5 - 6 | 695 | 91 | |
XM_017008379.1 | XP_016863868.1 | 5 - 6 | 690 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |