Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015231.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015231.hg.1 |
TC19001106.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900176074.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176074.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900176074.hg.1 |
TC1900009452.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900176074.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176074.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009452.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900176075.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176075.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176075.hg.1 |
NR_024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900176075.hg.1 |
TC1900009452.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900176075.hg.1 |
ENST00000597372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176075.hg.1 |
ENST00000600751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176075.hg.1 |
NR_024075.1 |
Refseq |