Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000428.2 | NP_000419.1 | 31 - 32 | 4970 | 76 | |
XM_011536765.1 | XP_011535067.1 | 29 - 30 | 4497 | 76 | ||
GenBank mRNA | AB209865.1 | - | 31 - 32 | 4220 | 76 | |
BC078659.1 | - | 32 - 33 | 4876 | 30343778 | 76 | |
S82451.1 | - | 31 - 32 | 4970 | 76 | ||
Z37976.1 | - | 31 - 32 | 4971 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |