Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000324913.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000324913.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000324913.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000324913.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000324913.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000409719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000409719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000409719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000409719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000409719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000409719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000409719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000414784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000414784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000454841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000454841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000454841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000454841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000454841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000454841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000469030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000469030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000469030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000469030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000489052.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000489052.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
ENST00000489052.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_001304353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_001304353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_001304353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_001304354.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_001304354.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_001304354.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_001304354.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_032309.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_032309.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_032309.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_032309.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008924.hg.1 |
NM_032309.3 |
Refseq |