Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000392517.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000392517.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000392517.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000392517.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000356348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000368564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000392517.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000392517.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000392517.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000392517.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
ENST00000413340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009240.hg.1 |
NM_002269.2 |
Refseq |