Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178229.4 | NP_839943.2 | 33 - 34 | 4265 | 74 | |
XM_011509198.2 | XP_011507500.1 | 33 - 34 | 4254 | 74 | ||
XM_011509200.2 | XP_011507502.1 | 22 - 23 | 3434 | 74 | ||
XM_011509201.2 | XP_011507503.1 | 21 - 22 | 2823 | 74 | ||
XM_017000317.1 | XP_016855806.1 | 33 - 34 | 4251 | 74 | ||
XM_017000318.1 | XP_016855807.1 | 31 - 32 | 4008 | 74 | ||
GenBank mRNA | AB105103.1 | - | 33 - 34 | 4201 | 74 | |
AK122653.1 | - | 19 - 20 | 2473 | 74 | ||
AY253300.1 | - | 33 - 34 | 4190 | 74 | ||
BC131536.1 | - | 33 - 34 | 4223 | 40082932 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |