Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135254.1 | NP_001128726.1 | 3 - 4 | 1045 | 55 | |
NM_002584.2 | NP_002575.1 | 3 - 4 | 1045 | 55 | ||
GenBank mRNA | DQ322591.1 | - | 3 - 4 | 530 | 55 | |
X96743.1 | - | 3 - 4 | 1046 | 55 | ||
Z35141.1 | - | 3 - 4 | 549 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |