Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEZ6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184773.1 | NP_001171702.1 | 12 - 13 | 2799 | 82 | |
NM_001184774.1 | NP_001171703.1 | 12 - 13 | 2799 | 82 | ||
NM_001184775.1 | NP_001171704.1 | 12 - 13 | 2799 | 82 | ||
NM_021115.4 | NP_066938.2 | 12 - 13 | 2799 | 82 | ||
XM_005261439.3 | XP_005261496.1 | 12 - 13 | 2775 | 82 | ||
XM_006724195.3 | XP_006724258.1 | 12 - 13 | 2775 | 82 | ||
GenBank mRNA | AB041736.1 | - | 12 - 13 | 2603 | 82 | |
AK294790.1 | - | 2 - 3 | 395 | 82 | ||
AL050253.1 | - | 11 - 12 | 2664 | 82 | ||
AY358405.1 | - | 12 - 13 | 2799 | 82 | ||
BC126115.1 | - | 12 - 13 | 2698 | 8991831 | 82 | |
BC143851.1 | - | 12 - 13 | 2698 | 9052367 | 82 | |
BC143854.1 | - | 12 - 13 | 2698 | 9052370 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |