Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEF2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193350.1 | NP_001180279.1 | 4 - 5 | 1054 | 119 | |
NM_001308002.1 | NP_001294931.1 | 3 - 4 | 542 | 119 | ||
NM_002397.4 | NP_002388.2 | 4 - 5 | 1057 | 119 | ||
XM_005248511.2 | XP_005248568.1 | 4 - 5 | 591 | 119 | ||
XM_006714619.2 | XP_006714682.1 | 5 - 6 | 913 | 119 | ||
XM_011543396.2 | XP_011541698.1 | 4 - 5 | 642 | 119 | ||
XM_011543400.1 | XP_011541702.1 | 4 - 5 | 829 | 119 | ||
XM_011543401.1 | XP_011541703.1 | 4 - 5 | 829 | 119 | ||
XM_017009476.1 | XP_016864965.1 | 4 - 5 | 890 | 119 | ||
XM_017009477.1 | XP_016864966.1 | 4 - 5 | 829 | 119 | ||
XM_017009479.1 | XP_016864968.1 | 4 - 5 | 890 | 119 | ||
XM_017009480.1 | XP_016864969.1 | 5 - 6 | 913 | 119 | ||
XM_017009481.1 | XP_016864970.1 | 4 - 5 | 591 | 119 | ||
XM_017009482.1 | XP_016864971.1 | 4 - 5 | 890 | 119 | ||
GenBank mRNA | AK312472.1 | - | 4 - 5 | 762 | 119 | |
BC026341.1 | - | 4 - 5 | 829 | 119 | ||
L08895.1 | - | 4 - 5 | 804 | 119 | ||
S57212.1 | - | 1063 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |