Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAAR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053278.1 | NP_444508.1 | 1 | 548 | 81 | |
GenBank mRNA | AY183468.1 | - | 1 | 548 | 81 | |
BC069166.1 | - | 1 | 548 | 7217002 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |