Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000263341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000416750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000416750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000416750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000416750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000416750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000418817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000418817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000418817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000418817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000418817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000418817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000418817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000418817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000432018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000432018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000432018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000432018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000432018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000432018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000477398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000477398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000477398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000477398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000487639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000487639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000487639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000487639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000487639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000487639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000487639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000491056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000496280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000496280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000496280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000496280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000496280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
ENST00000496280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013916.hg.1 |
NM_000576.2 |
Refseq |