Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015719.3 | NP_056534.2 | 24 - 25 | 2082 | 109 | |
XM_011528042.2 | XP_011526344.1 | 24 - 25 | 2140 | 109 | ||
GenBank mRNA | AB209175.1 | - | 22 - 23 | 1735 | 109 | |
AF177941.1 | - | 24 - 25 | 2082 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |